Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CBY1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113543
Description
CBY1 Polyclonal Antibody for Western Blot, IHC (P), ELISA
Beta-catenin is a transcriptional activator and oncoprotein involved in the development of several cancers. The protein encoded by this gene interacts directly with the C-terminal region of beta-catenin, inhibiting oncogenic beta-catenin-mediated transcriptional activation by competing with transcription factors for binding to beta-catenin. Two transcript variants encoding different isoforms have been found for this gene.
Specifications
| CBY1 | |
| Polyclonal | |
| Unconjugated | |
| CBY1 | |
| 1110014P06Rik; arb1; ARPP-binding protein; C22orf2; CBY; CBY1; chibby CTNNB1-mediated transcription inhibitor; chibby family member 1, beta catenin antagonist; chibby homolog 1; chibby homolog 1 (Drosophila); coiled-coil protein PIGEA-14; cytosolic leucine-rich protein; HRIHFB2025; HS508I15A; PGEA1; PGEA14; PIGEA14; PIGEA-14; PKD2 interactor, golgi and endoplasmic reticulum associated 1; PKD2 interactor, Golgi and endoplasmic reticulum-associated 1; polycystin-2 interactor, Golgi- and endoplasmic reticulum-associated protein, 14 kDa; protein chibby homolog 1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 25776, 73739 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 1.36 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9D1C2, Q9Y3M2 | |
| CBY1 | |
| Recombinant Human Protein chibby homolog 1 protein (1-126AA). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction