Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ CC 401 dihydrochloride
GREENER_CHOICE

Catalog No. 625810
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
62-581-0 10 mg
62-585-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 62-581-0 Supplier Tocris Bioscience™ Supplier No. 6258/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

High affinity JNK inhibitor; also inhibits HCMV replication

High affinity JNK inhibitor (Ki values are 25-50 nM). Inhibits JNK via competitive binding of the ATP-binding site of active, phosphorylated JNK. Exhibits > 40-fold selectivity for JNK over p38, ERK, IKK2, protein kinase C, Lck and ZAP70. Hepatoprotective. Also inhibits HCMV replication.

Chemical Identifiers

CAS 2250025-96-6

Specifications

Quantity 10 mg
Solubility Soluble to 100mM in water and to 100mM in DMSO
Formula Weight 461.39
Physical Form Solid
Purity >98%
Chemical Name or Material 3-[3-[2-(1-Piperidinyl)ethoxy]phenyl]-5-(1H-1,2,4-triazol-5-yl)-1H-indazole dihydrochloride
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.