Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DIPK2A Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA526926

Catalog No. PIPA526926


Only null left
Add to Cart

Description

Description

DIPK2A Polyclonal Antibody for Western Blot, IHC (P)

DIPK2A (CC058, 1190002N15Rik) may play a role in cardiomyocyte proliferation through paracrine signaling and activation of the PPI3K-AKT-CDK7 signaling cascade.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

DIPK2A
Polyclonal
Unconjugated
DIPK2A
1190002N15Rik; Ab2-095; C3orf58; cc058; chromosome 3 open reading frame 58; deleted in autism 1; deleted in autism protein 1; Deleted in autism protein 1 homolog; DIA1; Dipk2a; Divergent protein kinase domain 2A; Golgi protein GoPro49; Golgi Protein of 49 kDa; GoPro49; HASF; hypoxia and Akt induced stem cell factor; hypoxia and AKT-induced stem cell factor; RGD1309079; RIKEN cDNA 1190002N15 gene; similar to Ab2-095; UPF0672 protein C3orf58; UPF0672 protein C3orf58 homolog
Rabbit
Antigen affinity chromatography, Protein A
RUO
205428, 68861
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.4
Q3USZ8, Q8NDZ4
DIPK2A
KLH conjugated synthetic peptide between 159-188 amino acids from the central region of human CC058.
400 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.