Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CC2D1A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA568384
Description
Predicted to react with Mouse and Rat samples.
This gene encodes a transcriptional repressor that binds to a conserved 14-bp 5'-repressor element and regulates expression of the 5-hydroxytryptamine (serotonin) receptor 1A gene in neuronal cells. The DNA binding and transcriptional repressor activities of the protein are inhibited by calcium. A mutation in this gene results in nonsyndromic mental retardation-3.
Specifications
CC2D1A | |
Polyclonal | |
Unconjugated | |
CC2D1A | |
5-HT1A receptor; AKI1; Akt kinase-interacting protein 1; BC016188; CC2D1A; coiled-coil and C2 domain containing 1A; coiled-coil and C2 domain-containing protein 1A; Five prime repressor element under dual repression-binding protein 1; five' repressor element under dual repression binding protein 1; five repressor element under dual repression-binding protein 1; FLJ20241; FRE under dual repression-binding protein 1; Freud-1; Freud-1/Aki1; mental retardation, nonsyndromic, autosomal recessive, 3; MRT3; Putative NF-kappa-B-activating protein 023N; putative NFkB activating protein; RGD1306108; Tape | |
Rabbit | |
affinity chromatography | |
RUO | |
212139, 288908, 54862 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
1.4 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q66HA5, Q6P1N0, Q8K1A6 | |
CC2D1A | |
Synthetic peptide corresponding to residues near the C terminal of human CC2D1A. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction