Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CC2D1A Recombinant Rabbit Monoclonal Antibody (5Y6S6)
GREENER_CHOICE

Catalog No. PIMA542700
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA542700 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA542700 Supplier Invitrogen™ Supplier No. MA542700
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Positive test controls include: HeLa, U-251MG, Mouse testis, Mouse brain. The target is usually found in the following locations: Cytoplasm, Nucleus, centrosome, cytoskeleton, microtubule organizing center. Immunogen sequence: VLGTAQLKLD ALEIACEVRE ILEVLDGRRP TGGRLEVMVR IREPLTAQQL ETTTERWLVI DPVPAAVPTQ VAGPKGKAPP VPAPARESGN RSARPLHSLS VLAFDQERLE RKILALRQAR RPVPPEVAQQ YQDIMQRSQW QRAQLEQGGV GIRREYAAQL ERQLQFYTEA ARRLGNDGSR DAAKEALYRR NLVESELQRL RR.

This gene encodes a transcriptional repressor that binds to a conserved 14-bp 5'-repressor element and regulates expression of the 5-hydroxytryptamine (serotonin) receptor 1A gene in neuronal cells. The DNA binding and transcriptional repressor activities of the protein are inhibited by calcium. A mutation in this gene results in nonsyndromic mental retardation-3.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CC2D1A
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone 5Y6S6
Concentration 1.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CC2D1A
Gene Accession No. Q66HA5, Q6P1N0, Q8K1A6
Gene Alias 5-HT1A receptor; AKI1; Akt kinase-interacting protein 1; BC016188; CC2D1A; coiled-coil and C2 domain containing 1A; coiled-coil and C2 domain-containing protein 1A; Five prime repressor element under dual repression-binding protein 1; five' repressor element under dual repression binding protein 1; five repressor element under dual repression-binding protein 1; FLJ20241; FRE under dual repression-binding protein 1; Freud-1; Freud-1/Aki1; mental retardation, nonsyndromic, autosomal recessive, 3; MRT3; Putative NF-kappa-B-activating protein 023N; putative NFkB activating protein; RGD1306108; Tape
Gene Symbols CC2D1A
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 750-951 of human CC2D1A (Q6P1N0).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 212139, 288908, 54862
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.