Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CC2D1B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565098
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565098 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565098 Supplier Invitrogen™ Supplier No. PA565098
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RIGKRFGAVLE ALEKGQPVDL SAMPPAPEDL KPQQASQAPT APSVIPPAVE RVQPVMAPDV PATPVAPTES QTVLDALQQR LNKYREAGIQ ARS Highest antigen sequence identity to the following orthologs - mouse 77%, rat 76%.

Transcription factor that binds specifically to the DRE (dual repressor element) and represses HTR1A gene transcription in neuronal cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CC2D1B
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CC2D1B
Gene Accession No. Q5T0F9
Gene Alias A830039B04Rik; CC2D1B; coiled-coil and C2 domain containing 1B; coiled-coil and C2 domain-containing protein 1B; Five prime repressor element under dual repression-binding protein 2; FRE under dual repression-binding protein 2; Freud2; freud-2; KIAA1836; RGD1306630
Gene Symbols CC2D1B
Host Species Rabbit
Immunogen Recombinant Human CC2D1B. Recombinant protein control fragment (Product #RP-104631).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 200014
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.