Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CC2D2A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560621
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560621 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560621 Supplier Invitrogen™ Supplier No. PA560621
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SVTPNDQCPR AEVSRREDVK KRSVYLKVLF NNKEVSRTVS RPLGADFRVH FGQIFNLQIV NWPESLTLQV YETVGHSSPT LLAEVFLPIP ETTVVTGRAP TEEVEFSSN Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 89%.

This gene encodes a coiled-coil and calcium binding domain protein that appears to play a critical role in cilia formation. Mutations in this gene cause Meckel syndrome type 6, as well as Joubert syndrome type 9. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CC2D2A
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CC2D2A
Gene Accession No. Q9P2K1
Gene Alias 5730509K17Rik; b2b1035Clo; CC2D2A; coiled-coil and C2 domain containing 2A; coiled-coil and C2 domain-containing protein 2A; JBTS9; KIAA1345; MKS6
Gene Symbols CC2D2A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CC2D2A. Recombinant protein control fragment (Product #RP-100190).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57545
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.