Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CC2D2B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA583704
Description
Immunogen sequence: VPSSSPVVNQ RKLPKDMMPR ILEDEGFYIQ RKPEIYKKTC NKMENRLLKL EEGKCWFGES GEIMSLPTPI KQSWNF.
CC2D2B is a protein coding gene.
Specifications
CC2D2B | |
Polyclonal | |
Unconjugated | |
CC2D2B | |
bA248J23.4; C10orf130; C10orf131; CC2D2B; coiled-coil and C2 domain containing 2B; protein CC2D2B | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
387707 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6DHV5 | |
CC2D2B | |
Recombinant protein corresponding to Human CC2D2B. Recombinant protein control fragment (Product #RP-108167). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction