Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCAP Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989402
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9402 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9402 Supplier Bioss Supplier No. BS9859R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SDCCAG8 (serologically defined colon cancer antigen 8), also known as CCCAP (centrosomal colon cancer autoantigen protein), HSPC085 or NY-CO-8, is a 713 amino acid cytoplasmic protein that is expressed in thymus, prostate, testis, ovary, small intestine, colon, mucosa and renal cancer tumors. Existing as a homodimer, SDCCAG8 localizes to centrioles and interacts with oral-facial-digital syndrome 1 (ODF1), which is associated with nephronophthisis-related ciliopathies (NPHP-RC), a recessive disorder that is characterized by dysplasia or degeneration of the kidney, retina and cerebellum. SDCCAG8 exists as four alternatively spliced isoforms and is encoded by a gene located on humanc chromosome 1, which spans 260 million base pairs, contains over 3,000 genes and comprises nearly 8% of the human genome.

Specifications

Antigen CCAP
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene SDCCAG8
Gene Accession No. Q86SQ7
Gene Alias 2700048G21Rik; 5730470G24Rik; Antigen NY-CO-8; BBS16; CCCAP; CCCAP SLSN7; Centrosomal colon cancer autoantigen protein; hCCCAP; HSPC085; mCCCAP; NPHP10; NY-CO-8; SDCCAG8; Serologically defined colon cancer antigen 8; Serologically defined colon cancer antigen 8 homolog; si:dkey-60b15.1; slinky; SLSN7
Gene Symbols SDCCAG8
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from fruit fly Cardioactive Peptide.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10806
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.