Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCAR1 Antibody, Alexa Fluor™ 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB100258AF488
Description
CCAR1 Polyclonal antibody specifically detects CCAR1 in Human samples. It is validated for Western BlotSpecifications
CCAR1 | |
Polyclonal | |
Western Blot | |
CARP-1, CARP1MGC44628, Cell cycle and apoptosis regulatory protein 1, cell division cycle and apoptosis regulator 1, cell division cycle and apoptosis regulator protein 1, Death inducer with SAP domain, DIS, FLJ10590, RP11-437A18.1 | |
The immunogen recognized by this antibody maps to a region between residue 700 and 750 of human Cell-Cycle and Apoptosis Regulatory Protein 1 using the numbering given in entry NP_060707.2 (GeneID 55749). | |
0.1 mL | |
Apoptosis, Cell Cycle and Replication, DNA Repair | |
55749 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 488 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction