Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCAR1 Antibody, DyLight 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB500186UV
Description
CCAR1 Polyclonal antibody specifically detects CCAR1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
CCAR1 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
CARP-1, CARP1MGC44628, Cell cycle and apoptosis regulatory protein 1, cell division cycle and apoptosis regulator 1, cell division cycle and apoptosis regulator protein 1, Death inducer with SAP domain, DIS, FLJ10590, RP11-437A18.1 | |
A synthetic peptide, mapping to a region region between residues 1 and 50 of human Cell-Cycle and Apoptosis Regulatory Protein 1 using the numbering given in TrEMBL entry Q6X935 (GeneID 55749). | |
0.1 mL | |
Apoptosis, Cell Cycle and Replication, DNA Repair | |
55749 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
DyLight 350 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction