Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCAR1 Mouse anti-Human, MilliporeSigma™

Catalog No. MABE167125 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE1671-25 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE1671-25 Supplier MilliporeSigma Supplier No. MABE167125UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-CCAR1, clone 4B9.1 is a highly specific mouse monoclonal antibody, that targets Cell division cycle and apoptosis regulator protein 1 and has been tested in Western Blotting and Immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCAR1
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols CCAR1,CARP1,DIS
Host Species Mouse
Immunogen GST/His-tagged recombinant fragment corresponding to 173 amino acids from the N-terminal half of human Cell division cycle and apoptosis regulator protein 1 (CCAR1).
Purification Method Protein G Purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Test Specificity Clone 4B9.1 detects Cell division cycle and apoptosis regulator protein 1 (CCAR1) in human cells. It targets an epitope witihin 173 amino acids from the N-terminal half.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.