Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CCAR1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA552473
Description
Immunogen sequence: KDVEENVGLI VYNGAMVDVG SLLQKLEKSE KVRAEVEQKL QLLEEKTDED EKTILNLENS NKSLSGELRE VKKDLSQLQE NLKISENMNL QFENQMNKTI RNLSTVMDEI HTVLKKDNVK NED Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 94%.
IFI-44 (interferon-induced protein 44), also known as p44 or MTAP44 (microtubule-associated protein 44), is a 444 amino acid protein that localizes to the cytoplasm and, upon induction by IFN-betas, aggregates to form microtubular structures. Human IFI-44 shares 97% sequence similarity with its chimp counterpart, suggesting a conserved role between species. The gene encoding IFI-44 maps to human chromosome 1, which spans 260 million base pairs, contains over 3,000 genes and comprises nearly 8% of the human genome. Chromosome 1 houses a large number of disease-associated genes, including those that are involved in familial adenomatous polyposis, Stickler syndrome, Parkinson's disease, Gaucher disease, schizophrenia and Usher syndrome.
Specifications
CCAR1 | |
Polyclonal | |
Unconjugated | |
CCAR1 | |
2610511G16Rik; 9430036H15Rik; Carp1; CARP-1; Ccar1; Cell cycle and apoptosis regulatory protein 1; cell division cycle and apoptosis regulator 1; cell division cycle and apoptosis regulator protein 1; Death inducer with SAP domain; DIS; MGC44628; OTTHUMP00000019709; RGD1560358; RP11-437A18.1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
361849, 55749, 67500 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8CH18, Q8IX12 | |
CCAR1 | |
Recombinant protein corresponding to Human CCAR1. Recombinant protein control fragment (Product #RP-89830). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction