Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCAR2 Mouse anti-Human, Porcine, Clone: 2D10F8, Proteintech
SDP

Catalog No. 501736920 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6920 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6920 Supplier Proteintech Group Inc Supplier No. 664971IG150UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DBC1 is located within a chromosomal region that shows loss of heterozygosity in some bladder cancers. It contains a 5′ CpG island that may be a frequent target of hypermethylation, and it may undergo hypermethylation-based silencing in some bladder cancers.

Specifications

Antigen CCAR2
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 2D10F8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene CCAR2
Gene Accession No. Q8N163
Gene Alias CCAR2, cell cycle and apoptosis regulator 2, DBC 1, DBC.1, DBC1, KIAA1967, p30 DBC, Protein KIAA1967
Gene Symbols CCAR2
Host Species Mouse
Immunogen CCAR2 Fusion Protein Ag18374
Purification Method Protein A
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100155718, 57805
Target Species Human, Pig
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.