Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CCBE1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA559534
Description
Immunogen sequence: CECREGYIRE DDGKTCTRGD KYPNDTGHEK SENMVKAGTC CATCKEFYQM KQTVLQLKQK IALLPNNAAD LGKYITGDKV LASNTYLPGP PG Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 90%.
The gene encoding CCBE1 (Collagen and calcium-binding EGF domain-containing protein 1) is located on chromosome 18 in a region that is frequently found to be deleted in breast and prostate cancers. CCBE1 is a 406 amino acid secreted protein that contains two collagen-like domains and one calcium-binding epidermal growth factor (EGF-like) domain. In general, EGF-like domains include six cysteine residues that are usually involved in disulfide bonds. Calcium-binding EGF-like domains are subject to amino acid substitutions that disrupt their structure or calcium affinity, therefore altering protein function. Downregulation of the gene encoding CCBE1 is observed with high frequency in breast cancer, suggesting that loss of CCBE1 results in changes in cell adhesion and mobility due to its characterization as an extracellular protein. There are three named isoforms of CCBE1 which are produced as a result of alternative splicing events.
Specifications
CCBE1 | |
Polyclonal | |
Unconjugated | |
CCBE1 | |
4933426F18Rik; 9430093N24Rik; CCBE1; collagen and calcium binding EGF domains 1; collagen and calcium-binding EGF domain-containing protein 1; FLJ30681; fof; full of fluid protein; Full of fluid protein homolog; HKLLS1; KIAA1983; MGC50861; mKIAA1983; RGD1307670; UNQ1921/PRO4395 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
147372 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6UXH8 | |
CCBE1 | |
Recombinant protein corresponding to Human CCBE1. Recombinant protein control fragment (Product #RP-98285). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction