Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CCBE1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA599806
Description
Antibody detects endogenous levels of total CCBE1.
The gene encoding CCBE1 (Collagen and calcium-binding EGF domain-containing protein 1) is located on chromosome 18 in a region that is frequently found to be deleted in breast and prostate cancers. CCBE1 is a 406 amino acid secreted protein that contains two collagen-like domains and one calcium-binding epidermal growth factor (EGF-like) domain. In general, EGF-like domains include six cysteine residues that are usually involved in disulfide bonds. Calcium-binding EGF-like domains are subject to amino acid substitutions that disrupt their structure or calcium affinity, therefore altering protein function. Downregulation of the gene encoding CCBE1 is observed with high frequency in breast cancer, suggesting that loss of CCBE1 results in changes in cell adhesion and mobility due to its characterization as an extracellular protein. There are three named isoforms of CCBE1 which are produced as a result of alternative splicing events.
Specifications
CCBE1 | |
Polyclonal | |
Unconjugated | |
CCBE1 | |
4933426F18Rik; 9430093N24Rik; CCBE1; collagen and calcium binding EGF domains 1; collagen and calcium-binding EGF domain-containing protein 1; FLJ30681; fof; full of fluid protein; Full of fluid protein homolog; HKLLS1; KIAA1983; MGC50861; mKIAA1983; RGD1307670; UNQ1921/PRO4395 | |
Rabbit | |
Affinity chromatography | |
RUO | |
147372, 320924 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q3MI99, Q6UXH8 | |
CCBE1 | |
A synthesized peptide derived from human CCBE1(Accession Q6UXH8), corresponding to amino acid residues G155-F205. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction