Learn More
CCBE1, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA559541
Description
The gene encoding CCBE1 (Collagen and calcium-binding EGF domain-containing protein 1) is located on chromosome 18 in a region that is frequently found to be deleted in breast and prostate cancers. CCBE1 is a 406 amino acid secreted protein that contains two collagen-like domains and one calcium-binding epidermal growth factor (EGF-like) domain. In general, EGF-like domains include six cysteine residues that are usually involved in disulfide bonds. Calcium-binding EGF-like domains are subject to amino acid substitutions that disrupt their structure or calcium affinity, therefore altering protein function. Downregulation of the gene encoding CCBE1 is observed with high frequency in breast cancer, suggesting that loss of CCBE1 results in changes in cell adhesion and mobility due to its characterization as an extracellular protein. There are three named isoforms of CCBE1 which are produced as a result of alternative splicing events.
Immunogen sequence: PGSFDFLLLM LADIRNDITE LQEKVFGHRT HSSAEEFPLP QEFPSYPEAM DLGSGDDHPR RTETRDLRAHighest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 81%.
Specifications
CCBE1 | |
Polyclonal | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6UXH8, Q3MI99 | |
CCBE1 | |
Recombinant protein corresponding to Human CCBE1 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot | |
Unconjugated | |
CCBE1 | |
Collagen and calcium-binding EGF domain-containing protein 1, Full of fluid protein homolog, KIAA1983, UNQ1921/PRO4395 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
147372, 320924, 361341 | |
Store at 4°C short term. Store at −20°C long term. Avoid freeze/thaw cycles. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Research Use Only