Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCBL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100131
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100131 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100131 Supplier Invitrogen™ Supplier No. PA5100131
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total CCBL1/KAT1.

Kynurenine aminotransferases KAT I, KAT II, and KAT III belong to the class-I pyridoxal-phosphate-dependent aminotransferase family. KAT I is a cytoplasmic protein involved in glutamine catabolism. KAT I functions in the catalysis of the transamination of L-kinurenine to form kynurenic acid, a neuroprotective and anticonvulsant metabolite of tryptophan. Kynurenic acid is involved in synaptic transmission and has been implicated in a number of neurological disorders including schizophrenia and Huntington's disease. KAT I also functions in the metabolism of cysteine conjugates in some halogenated alkenes and alkanes to form reactive metabolites. KAT I has three isoforms. Isoform 1is the full length protein, isoform 2 lacks amino acids 68-117 and isoform 3 lacks amino acids 251-422. Based on sequence similarity, KAT I is thought to function as a homodimer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCBL1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene KYAT1
Gene Accession No. Q08415, Q16773, Q8BTY1
Gene Alias 2010009K05Rik; AI182306; beta-lysase, kidney; CCBL1; cysteine conjugate beta lyase 1; cysteine conjugate-beta lyase 1; cysteine conjugate-beta lyase, cytoplasmic; cysteine conjugate-beta lyase; cytoplasmic (glutamine transaminase K, kyneurenine aminotransferase); cysteine-S-conjugate beta-lyase; cytoplasmic (glutamine transaminase K, kyneurenine aminotransferase); glutamine transaminase K; glutamine-phenylpyruvate aminotransferase; Glutamine--phenylpyruvate transaminase; Gtk; Kat; KAT1; KATI; KYAT1; kyneurenine aminotransferase; kynurenine aminotransferase 1; Kynurenine aminotransferase I; kynurenine aminotransferase/glutamine transaminase K; kynurenine--oxoglutarate transaminase 1; Kynurenine--oxoglutarate transaminase 1, mitochondrial; kynurenine--oxoglutarate transaminase I
Gene Symbols KYAT1
Host Species Rabbit
Immunogen A synthesized peptide derived from human KYAT1(Accession Q16773), corresponding to amino acid residues S326-L376.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 311844, 70266, 883
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.