Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCBL2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $646.00
Specifications
Antigen | CCBL2 |
---|---|
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
CCBL2 Polyclonal specifically detects CCBL2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
CCBL2 | |
Polyclonal | |
Rabbit | |
Human, Mouse, Rat | |
cysteine conjugate-beta lyase 2, Cysteine-S-conjugate beta-lyase 2, DKFZp547N1117, EC 2.6.1.63, EC 2.6.1.7, EC 4.4.1.13, KAT3DKFZp667D0223, KATIII, Kynurenine aminotransferase III, Kynurenine--glyoxylate transaminase, kynurenine-oxoglutarate transaminase 3, kynurenine--oxoglutarate transaminase 3, Kynurenine--oxoglutarate transaminase III, MGC9398, RBM1, RP11-82K18.3, RP4-531M19.2 | |
CCBL2 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
56267 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:PDGGYFIIADVSLLDPDLSDMKNNEPYDYKFVKWMTKHKKLSAIPVSAFCNSETKSQFEKFVRFCFIKKDSTLDAAEEIIKAWSVQKS | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title