Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCBL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541727
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541727 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541727 Supplier Invitrogen™ Supplier No. PA541727
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: SGRAKFLKTI SSSKILGFST SAKMSLKFTN AKRIEGLDSN VWIEFTKLAA Sequence homology: Cow: 92%; Dog: 93%; Guinea Pig: 85%; Horse: 93%; Human: 100%; Mouse: 93%; Pig: 93%; Rabbit: 86%; Rat: 91%; Zebrafish: 91%.

RP11-82K18.3 is an aminotransferase that transaminates kynurenine to form kynurenic acid. Kynurenic acid is a metabolite of tryptophan.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCBL2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene KYAT3
Gene Accession No. Q6YP21
Gene Alias Ccbl2; cysteine conjugate beta lyase 2; cysteine conjugate-beta lyase 2; cysteine-S-conjugate beta-lyase 2; KAT3; KATIII; Kyat3; Kynurenine aminotransferase 3; kynurenine aminotransferase III; Kynurenine--glyoxylate transaminase; kynurenine-oxoglutarate transaminase 3; kynurenine--oxoglutarate transaminase 3; Kynurenine--oxoglutarate transaminase 3-like protein; kynurenine--oxoglutarate transaminase III; RP11-82K18.3; Unknown (protein for MGC:140498); zgc:63929
Gene Symbols KYAT3
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human RP11-82K18.3 (aa 12-61).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56267
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.