Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCD33 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA524486

Catalog No. PIPA524486


Only null left
Add to Cart

Description

Description

CCD33 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY
Specifications

Specifications

CCD33
Polyclonal
PBS with 0.09% sodium azide; pH 7.4
Q8N5R6
CCDC33
KLH conjugated synthetic peptide between 873-901 amino acids from the C-terminal region of human CCD33.
400 μL
Primary
Human
Antibody
IgG
Western Blot
Unconjugated
CCDC33
Cancer/testis antigen 61; CCDC33; coiled-coil domain containing 33; coiled-coil domain-containing protein 33; CT61; HP11097; hypothetical protein HP11097
Rabbit
Antigen Affinity Chromatography, Protein A
RUO
80125
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.