Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CCDC92 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA566093
Description
Immunogen sequence: SPHFSSYDEGP LDVSMAATNL ENQLHSAQKN LLFLQREHAS TLKGLHSEIR RLQQHCTDLT YELTVKSSEQ TGDGTSKSSE LK Highest antigen sequence identity to the following orthologs - mouse 71%, rat 72%.
CCDC92 (Coiled-Coil Domain Containing 92) is a Protein Coding gene.
Specifications
CCDC92 | |
Polyclonal | |
Unconjugated | |
Ccdc92 | |
AW050178; Ccdc92; coiled-coil domain containing 92; coiled-coil domain-containing protein 92; D5Bwg0834e; limkain beta 2; limkain beta-2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
80212 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q53HC0 | |
Ccdc92 | |
Recombinant Human CCD92. Recombinant protein control fragment (Product #RP-106790). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction