Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCDC102A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559369
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559369 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559369 Supplier Invitrogen™ Supplier No. PA559369
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LPWEDTAATE EEASKLTALR LRLDESQKVL LKEREDKLAL SRNIEKLEGE LSQWKIKYEE LSKTKQEMLK QL Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 94%.

CCDC102A gene ontology (GO) annotation includes motor activity and myosin complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCDC102A
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CCDC102A
Gene Accession No. Q96A19
Gene Alias BC027663; CCDC102A; coiled-coil domain containing 102A; coiled-coil domain-containing protein 102A; Gm586
Gene Symbols CCDC102A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CCDC102A. Recombinant protein control fragment (Product #RP-98200).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 92922
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.