Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCDC102B Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP132619
Description
CCDC102B Polyclonal specifically detects CCDC102B in Human samples. It is validated for Western Blot.Specifications
CCDC102B | |
Polyclonal | |
Unconjugated | |
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal | |
CCDC102B | |
Recombinant protein encompassing a sequence within the center region of human CCDC102B. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
79839 | |
Human | |
IgG |
Western Blot | |
0.97mg/mL | |
Western Blot 1:500-1:3000 | |
ACY1L, aminoacylase 1-like, AN, C18orf14, chromosome 18 open reading frame 14, coiled-coil domain containing 102B, coiled-coil domain-containing protein 102B, DKFZp434K1426, DKFZp686I08254, FLJ23594, HsT1731, MGC161726, MGC161728 | |
Rabbit | |
60 kDa | |
0.1 mL | |
Primary | |
Tested in human. Not yet tested in any other species. | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction