Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCDC103 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113107
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113107 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113107 Supplier Invitrogen™ Supplier No. PA5113107
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CCDC103 Polyclonal Antibody for Western Blot, ELISA

CCDC103 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCDC103
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene CCDC103
Gene Accession No. Q8IW40, Q9D9P2
Gene Alias 1700039D13Rik; Ccdc103; CILD17; coiled-coil domain containing 103; coiled-coil domain-containing protein 103; coiled-coil domain-containing protein 103-like; PR46b; RGD1562370; SMH
Gene Symbols CCDC103
Host Species Rabbit
Immunogen Recombinant Human Coiled-coil domain-containing protein 103 protein (1-242AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 388389, 73293
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.