Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCDC104 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553541
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553541 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553541 Supplier Invitrogen™ Supplier No. PA553541
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DEEESKLTYT EIHQEYKELV EKLLEGYLKE IGINEDQFQE ACTSPLAKTH TSQAILQPVL AAEDFTIFKA MMVQKNIEMQ LQAIRIIQER NGVLPDCLTD GSDVVSDLEH EEMKILREVL RKSKEEYDQE EERKRK Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.

The CCDC104 (Coiled-coil domain-containing protein 104) protein, whose function is unknown, is expressed in various human tissues, including the brain, and is localized mainly to the nucleus, but is also found in the cytoplasm. It may act as an effector for ARL3. This protein has 2 isoforms produced by alternative splicing.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCDC104
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CFAP36
Gene Accession No. Q96G28
Gene Alias 2300003H10; 4931428D14Rik; BARTL1; CCDC104; CFAP36; cilia and flagella associated protein 36; cilia- and flagella-associated protein 36; coiled-coil domain containing 104; coiled-coil domain-containing protein 104; RGD1311732; UNQ163/PRO189
Gene Symbols CFAP36
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CCDC104. Recombinant protein control fragment (Product #RP-89856).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 112942
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.