Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCDC106 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113331
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113331 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113331 Supplier Invitrogen™ Supplier No. PA5113331
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CCDC106 Polyclonal Antibody for Western Blot, IHC (P), ELISA

The coiled-coil domain is a common protein motif that is often involved in protein oligomerization and is found in proteins such as transcription factors and intermediate filaments. CCDC106 was initially identified as a p53-interacting protein by yeast two-hybrid screening. Other experiments demonstrated that CCDC106 co-localizes and interacts with p53 in the nucleus, inhibiting the transcriptional activity of p53 and stimulating p53 protein degradation, indicating that at least one of the functions of CCDC106 is acting as a negative regulator of p53.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCDC106
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene CCDC106
Gene Accession No. Q3ULM0, Q9BWC9
Gene Alias BC018462; CCDC106; coiled-coil domain containing 106; coiled-coil domain-containing protein 106; HSU79303; MNCb-2071; protein predicted by clone 23882; RGD1566222; ZNF581
Gene Symbols CCDC106
Host Species Rabbit
Immunogen Recombinant Human Coiled-coil domain-containing protein 106 protein (1-280AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 232821, 29903
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.