Learn More
Invitrogen™ CCDC106 Polyclonal Antibody

Description
A suggested positive control is human brain tissue lysate. PA5-21003 can be used with blocking peptide PEP-1117.
Specifications
Specifications
| Antigen | CCDC106 |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | CCDC106 |
| Gene Accession No. | Q3ULM0, Q9BWC9 |
| Gene Alias | BC018462; CCDC106; coiled-coil domain containing 106; coiled-coil domain-containing protein 106; HSU79303; MNCb-2071; protein predicted by clone 23882; RGD1566222; ZNF581 |
| Gene Symbols | CCDC106 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.