Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CCDC107 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA531600
Description
Recommended positive controls: A549. Predicted reactivity: Human (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a membrane protein which contains a coiled-coil domain in the central region. Multiple alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications
CCDC107 | |
Polyclonal | |
Unconjugated | |
CCDC107 | |
CCDC107; coiled-coil domain containing 107; coiled-coil domain-containing protein 107; PSEC0222 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
203260 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q8WV48 | |
CCDC107 | |
Recombinant protein fragment corresponding to a region within amino acids 30 and 267 of Human CCDC107. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction