Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCDC107 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565619
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565619 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565619 Supplier Invitrogen™ Supplier No. PA565619
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DRANPDLRAHP GNAAHPGSGA TEPRRRPPLK DQRERT Highest antigen sequence identity to the following orthologs - mouse 54%, rat 65%.

This gene encodes a membrane protein which contains a coiled-coil domain in the central region. Multiple alternatively spliced transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCDC107
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CCDC107
Gene Accession No. Q8WV48
Gene Alias CCDC107; coiled-coil domain containing 107; coiled-coil domain-containing protein 107; PSEC0222
Gene Symbols CCDC107
Host Species Rabbit
Immunogen Recombinant Human CCDC107. Recombinant protein control fragment (Product #RP-104902).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 203260
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.