Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCDC108 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113046
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113046 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113046 Supplier Invitrogen™ Supplier No. PA5113046
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CCDC108 Polyclonal Antibody for ELISA, ICC/IF, IHC (P), Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen CCDC108
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene CFAP65
Gene Accession No. Q6ZU64
Gene Alias CCDC108; CFAP65; cilia and flagella associated protein 65; Cilia- and flagella-associated protein 65; coiled-coil domain containing 108; coiled-coil domain-containing protein 108
Gene Symbols CFAP65
Host Species Rabbit
Immunogen Recombinant Human Cilia- and flagella-associated protein 65 protein (2-164AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 255101
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.