Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CCDC108 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA563075
Description
Immunogen sequence: TVDVPIHILG WNSALIHFQG VGYNPHMMGD TAPFHNISSW DNSSIHSRLV VPGQNVFLSQ SHISLGNIPV QSKCSRLLFL NNISKNEEIA FSWQPS Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 80%.

Specifications
CCDC108 | |
Polyclonal | |
Unconjugated | |
CFAP65 | |
CCDC108; CFAP65; cilia and flagella associated protein 65; Cilia- and flagella-associated protein 65; coiled-coil domain containing 108; coiled-coil domain-containing protein 108 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
255101 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6ZU64 | |
CFAP65 | |
Recombinant protein corresponding to Human CCDC108. Recombinant protein control fragment (Product #RP-101542). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction