Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCDC109B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561817
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561817 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561817 Supplier Invitrogen™ Supplier No. PA561817
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SNEHTAEMEH MKSLVHRLFT ILHLEESQKK REHHLLEKID HLKEQLQPLE QVKAGIEAHS E Highest antigen sequence identity to the following orthologs: Mouse - 75%, Rat - 74%.

Negatively regulates the activity of MCU, the mitochondrial inner membrane calcium uniporter, and thereby modulates calcium uptake into the mitochondrion. Does not form functional calcium channels by itself. Mitochondrial calcium homeostasis plays key roles in cellular physiology and regulates cell bioenergetics, cytoplasmic calcium signals and activation of cell death pathways.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCDC109B
Applications Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MCUB
Gene Accession No. Q9NWR8
Gene Alias calcium uniporter regulatory subunit MCUb, mitochondrial; CCDC109B; coiled-coil domain containing 109B; coiled-coil domain-containing protein 109B; MCUB; mitochondrial calcium uniporter dominant negative beta subunit; mitochondrial calcium uniporter regulatory subunit MCUb
Gene Symbols MCUB
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CCDC109B. Recombinant protein control fragment (Product #RP-101087).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55013
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.