Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCDC11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA544354
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA544354 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA544354 Supplier Invitrogen™ Supplier No. PA544354
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: YSQRFGTVQR EVKGPTPKVV IVRSKPPKGQ GAEHHLERIR RSHQKHNAIL Sequence homology: Cow: 93%; Dog: 79%; Guinea Pig: 77%; Horse: 85%; Human: 100%; Pig: 85%.

This gene belongs to the CFAP53 family. It was found to be differentially expressed by the ciliated cells of frog epidermis and in skin fibroblasts from human. Mutations in this gene are associated with visceral heterotaxy-6, which implicates this gene in determination of left-right asymmetric patterning. [provided by RefSeq, Aug 2015].
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCDC11
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene CFAP53
Gene Accession No. Q96M91
Gene Alias 4933415I03Rik; Ccdc11; Cfap53; cilia and flagella associated protein 53; cilia- and flagella-associated protein 53; coiled-coil domain containing 11; coiled-coil domain-containing protein 11; HTX6; mitogen-activated protein kinase kinase kinase kinase 4-like; RGD1306734
Gene Symbols CFAP53
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human CCDC11 (aa 2-51).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 220136
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.