Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCDC111 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144234
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144234 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144234 Supplier Invitrogen™ Supplier No. PA5144234
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CCDC111 Polyclonal Antibody for Western Blot, IHC (P), ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen CCDC111
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300, 0.02% sodium azide; pH 7.4
Gene PRIMPOL
Gene Accession No. Q96LW4
Gene Alias CCDC111; coiled-coil domain containing 111; coiled-coil domain-containing protein 111; DNA-directed primase/polymerase protein; hPrimpol1; MYP22; primase and DNA directed polymerase; primase and polymerase (DNA-directed); PRIMPOL
Gene Symbols PRIMPOL
Host Species Rabbit
Immunogen Recombinant Human DNA-directed primase/polymerase protein (305-537AA).
Purification Method Antigen Affinity Chromatography, Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 201973
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.