Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCDC112 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560930
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560930 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560930 Supplier Invitrogen™ Supplier No. PA560930
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NVKKLQHQLK DVKPTPDFVE KLREMMEEIE NAINTFKEEQ RLIYEELIKE EKTTNNELSA ISRKIDTWAL GNSETEKAFR AISSKVPVDK VTPSTLPE Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 91%.

CCDC112 (Coiled-Coil Domain Containing 112) is a Protein Coding gene and its exact function is unknown. An important paralog of this gene is CCDC148.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCDC112
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CCDC112
Gene Accession No. A0AUP1, Q8NEF3
Gene Alias 8430438M01Rik; AW108467; CCDC112; coiled-coil domain containing 112; coiled-coil domain-containing protein 112; MBC1; mutated in bladder cancer 1; mutated in bladder cancer protein 1; RGD1561942
Gene Symbols CCDC112
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CCDC112. Recombinant protein control fragment (Product #RP-100202).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 153733, 240261, 498858
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.