Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CCDC112 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA562223
Description
Immunogen sequence: DGCFSTSGGI RPFHLQNWKQ KVNQTKKAEF VRTAEKFKNQ VINMEKDKHS HFYNQKSDFR IEHSMLEELE NKLIHS Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 76%.
CCDC112 (Coiled-Coil Domain Containing 112) is a Protein Coding gene and its exact function is unknown. An important paralog of this gene is CCDC148.
Specifications
CCDC112 | |
Polyclonal | |
Unconjugated | |
CCDC112 | |
8430438M01Rik; AW108467; CCDC112; coiled-coil domain containing 112; coiled-coil domain-containing protein 112; MBC1; mutated in bladder cancer 1; mutated in bladder cancer protein 1; RGD1561942 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
153733 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8NEF3 | |
CCDC112 | |
Recombinant protein corresponding to Human CCDC112. Recombinant protein control fragment (Product #RP-103715). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction