Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCDC112 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598620
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598620 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598620 Supplier Invitrogen™ Supplier No. PA598620
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CCDC112 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

CCDC112 (Coiled-Coil Domain Containing 112) is a Protein Coding gene and its exact function is unknown. An important paralog of this gene is CCDC148.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCDC112
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 4 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene CCDC112
Gene Accession No. Q8NEF3
Gene Alias 8430438M01Rik; AW108467; CCDC112; coiled-coil domain containing 112; coiled-coil domain-containing protein 112; MBC1; mutated in bladder cancer 1; mutated in bladder cancer protein 1; RGD1561942
Gene Symbols CCDC112
Host Species Rabbit
Immunogen Recombinant Human Coiled-coil domain-containing protein 112 protein (201-446AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 153733
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.