Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCDC114 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560012
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560012 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560012 Supplier Invitrogen™ Supplier No. PA560012
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SKDDQHLLQE QQQKVLQQRM DKVHSEAERL EARFQDVRGQ LEKLKADIQL LFTKAHCDSS MIDDLLGVKT SMGDRDMGLF LSLIEKRLVE Highest antigen sequence identity to the following orthologs: Mouse - 68%, Rat - 70%.

The CCDC114 gene, located on chromosome 19, encodes a protein that is a key component of the structure and function of cilia, which are hair-like organelles present on the surface of many cell types. Structurally, CCDC114 belongs to the coiled-coil domain-containing family, which plays crucial roles in cellular organizations and motility. Functionally, CCDC114 is involved in forming the outer dynein arm of cilia, essential for their motility. Mutations in CCDC114 can lead to defects in ciliary structure, resulting in primary ciliary dyskinesia (PCD), a condition characterized by chronic respiratory tract infections, abnormal organ positioning, and other symptoms. Specifically, mutations like c.742G>A and c.584T>C have been identified to cause variant forms of PCD, as well as situs inversus, which is a complete mirror image reversal of the body's internal organ arrangement. Genetic investigations have shown that these mutations can significantly disrupt the function of cilia, confirming CCDC114's critical role in human ciliopathies.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCDC114
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.7 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CCDC114
Gene Accession No. Q96M63
Gene Alias Ccdc114; CILD20; coiled-coil domain containing 114; coiled-coil domain-containing protein 114; RGD1308141
Gene Symbols CCDC114
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CCDC114. Recombinant protein control fragment (Product #RP-98955).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 93233
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.