Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CCDC115 Monoclonal Antibody (4E9)

Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA524495
Description
Peptide Sequence: MGAKSVGPLQ YASHMEPQVC LHASEAQEGL QKFKVVRAGV HAPEEVGPRE AGLRRRKGPT KTPEPESSEA PQDPLNWFGI LVPHSLRQAQ ASFRDGLQLA ADIASLQNRI DWGRSQLRGL QEKLKQLEPG AA.
The CCDC115 gene encodes a protein known as coiled-coil domain-containing protein 115, which is involved in cellular processes related to membrane trafficking and Golgi apparatus function. CCDC115 is integral to maintaining Golgi homeostasis and is implicated in the functioning of the vacuolar H+-ATPase (V-ATPase) proton pump, critical for acidifying intracellular compartments. Mutations in CCDC115 have been associated with a genetic condition known as congenital disorder of glycosylation type IIo (CDG-IIo), which leads to defects in protein glycosylation. This disorder results in a spectrum of symptoms, including developmental delays, neurological impairments, and multi-systemic dysfunction due to improper glycosylation of proteins, impacting their stability and function. Research indicates that CCDC115's activity in ion transport and vacuolar acidification is essential for Golgi apparatus function and overall cellular homeostasis. Investigations into CCDC115 are crucial for understanding its role in glycosylation-related disorders and for developing potential therapeutic strategies to manage these conditions.
Specifications
CCDC115 | |
Monoclonal | |
0.5 mg/mL | |
PBS with no preservative; pH 7.4 | |
Q96NT0 | |
CCDC115 | |
CCDC115 (AAH06429.1, 1 a.a. ~ 132 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b κ |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
4E9 | |
Unconjugated | |
CCDC115 | |
2310061I09Rik; Ccdc115; Ccp1; CDG2O; coiled-coil domain containing 115; coiled-coil domain-containing protein 115; Coiled-coil protein 1; RGD1304653 | |
Mouse | |
Protein A | |
RUO | |
84317 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction