Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCDC115 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556987
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556987 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556987 Supplier Invitrogen™ Supplier No. PA556987
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PREAGLRRRK GPTKTPEPES SEAPQDPLNW FGILVPHSLR QAQASFRDGL QLAADIASLQ NRIDWGRSQL RGLQEKLKQL EP Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 84%.

The CCDC115 gene encodes a protein known as coiled-coil domain-containing protein 115, which is involved in cellular processes related to membrane trafficking and Golgi apparatus function. CCDC115 is integral to maintaining Golgi homeostasis and is implicated in the functioning of the vacuolar H+-ATPase (V-ATPase) proton pump, critical for acidifying intracellular compartments. Mutations in CCDC115 have been associated with a genetic condition known as congenital disorder of glycosylation type IIo (CDG-IIo), which leads to defects in protein glycosylation. This disorder results in a spectrum of symptoms, including developmental delays, neurological impairments, and multi-systemic dysfunction due to improper glycosylation of proteins, impacting their stability and function. Research indicates that CCDC115's activity in ion transport and vacuolar acidification is essential for Golgi apparatus function and overall cellular homeostasis. Investigations into CCDC115 are crucial for understanding its role in glycosylation-related disorders and for developing potential therapeutic strategies to manage these conditions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCDC115
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CCDC115
Gene Accession No. Q96NT0
Gene Alias 2310061I09Rik; Ccdc115; Ccp1; CDG2O; coiled-coil domain containing 115; coiled-coil domain-containing protein 115; Coiled-coil protein 1; RGD1304653
Gene Symbols CCDC115
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CCDC115. Recombinant protein control fragment (Product #RP-96090).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84317
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.