Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCDC116 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543957
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543957 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543957 Supplier Invitrogen™ Supplier No. PA543957
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: VQQEPATHTA QDQATEPCRS LYTNLPASRQ LSPLEPKLYM SACTGMGSSP Sequence homology: Human: 100%.

The function of this protein remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCDC116
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene CCDC116
Gene Accession No. Q8IYX3
Gene Alias 4930432J16Rik; Ccdc116; coiled-coil domain containing 116; coiled-coil domain-containing protein 116; Sdf2l1; stromal cell-derived factor 2-like 1
Gene Symbols CCDC116
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human CCDC116 (aa 526-575).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 164592
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.