Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCDC117 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
| Antigen | CCDC117 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
CCDC117 Polyclonal specifically detects CCDC117 in Human samples. It is validated for Western Blot.Specifications
| CCDC117 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| coiled-coil domain containing 117, coiled-coil domain-containing protein 117, dJ366L4.1, FLJ33814 | |
| CCDC117 | |
| IgG | |
| 30 kDa |
| Western Blot | |
| Unconjugated | |
| RUO | |
| NP_775781 | |
| 150275 | |
| Synthetic peptide directed towards the middle region of human CCDC117The immunogen for this antibody is CCDC117. Peptide sequence DTMKTGLKREFDEVFTKKMIESMSRPSMELVLWKPLPELLSDKPKPSSNT. | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title