Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCDC117 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116200
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116200 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116200 Supplier Invitrogen™ Supplier No. PA5116200
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CCDC117 Polyclonal Antibody for Western Blot, IHC (P)

Facilitates DNA repair, cell cycle progression, and cell proliferation through its interaction with CIAO2B.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCDC117
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene CCDC117
Gene Accession No. Q8IWD4
Gene Alias 1110004K02Rik; 1700026O03Rik; AU018638; AU042822; AV173073; BC018601; CCDC117; coiled-coil domain containing 117; coiled-coil domain-containing protein 117; dJ366L4.1
Gene Symbols CCDC117
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 143-171 amino acids from the C-terminal region of human CCDC117.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 150275
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.