Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCDC12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563350
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563350 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563350 Supplier Invitrogen™ Supplier No. PA563350
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EALRRKERLK ALREKTGRKD KEDGEPKTKH LREEEEEGEK HRELRLRNYV PEDEDLKKRR VPQAKPVAVE EKVKEQLEAA K Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 91%.

The exact function of this protein remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCDC12
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CCDC12
Gene Accession No. Q8WUD4
Gene Alias 2700094L05Rik; C76605; CCDC12; coiled-coil domain containing 12; coiled-coil domain-containing protein 12; RGD1306081
Gene Symbols CCDC12
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CCDC12. Recombinant protein control fragment (Product #RP-100179).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 151903
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.