Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CCDC120 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA548562
Description
CCDC120 Polyclonal Antibody for Western Blot
This gene encodes a protein that contains a coiled-coil domain. Several alternatively spliced transcript variants encoding multiple isoforms have been found for this gene. Required for neurite growth. Localizes CYTH2 to vesicles to allow its transport along neurites, and subsequent ARF6 activation and neurite growth.
Specifications
CCDC120 | |
Polyclonal | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q96HB5 | |
CCDC120 | |
KLH conjugated synthetic peptide between 59-88 amino acids from the N-terminal region of human CCDC120. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
CCDC120 | |
AA030924; CCDC120; coiled-coil domain containing 120; coiled-coil domain-containing protein 120; DXImx50e; JM11; RGD1561596; Sfc25 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
90060 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction