Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCDC120 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP310735100UL
Description
CCDC120 Polyclonal specifically detects CCDC120 in Human samples. It is validated for Western Blot.Specifications
CCDC120 | |
Polyclonal | |
Western Blot 1.0 ug/ml | |
coiled-coil domain containing 120, coiled-coil domain-containing protein 120, JM11 | |
The immunogen is a synthetic peptide directed towards the C-terminal region of Human CCDC120 (NP_296375). Peptide sequence SGSQDSQMGFPRADPASDRASLFVARTRRSNSSEALLVDRAAGGGAGSPP | |
100 μg | |
Primary | |
Human | |
Purified |
Western Blot | |
Unconjugated | |
PBS buffer, 2% sucrose | |
Rabbit | |
Affinity purified | |
RUO | |
90060 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction