Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCDC124 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589827
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589827 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589827 Supplier Invitrogen™ Supplier No. PA589827
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MPKKFQGENT KSAAARARRA EAKAAADAKK QKELEDAYWK DDDKHVMRKE QRKEEKEKRR LDQLERKKET QRLLEEEDSK LKGGKAPRVA TSSKVTRAQI EDTLRRDHQL REAPDTAEKA KSHLEVPLEE NVNRRVLEEG SVEARTIEDA IAVLSVAEEA ADRHPERRMR AAFTAFEEAQ LPRLKQENPN MRLSQLKQLL KKEWLRSPDN PMNQRAVPFN APK; Positive Samples: A-549, HeLa, U-87MG, Mouse pancreas, Mouse thymus, Mouse brain, Mouse spleen, Rat pancreas, Rat thymus; Cellular Location: Cytoplasm, Midbody, centrosome, cytoskeleton, microtubule organizing center.

Required for proper progression of late cytokinetic stages.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCDC124
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 5.04 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Ccdc124
Gene Accession No. Q96CT7, Q9D8X2
Gene Alias 1810023B24Rik; Ccdc124; coiled-coil domain containing 124; coiled-coil domain-containing protein 124; hypothetical protein LOC510778; RGD1310931
Gene Symbols Ccdc124
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 115098, 234388, 290642
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.