Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCDC124 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA546311
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA546311 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA546311 Supplier Invitrogen™ Supplier No. PA546311
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: VMRKEQRKEE KEKRRLDQLE RKKETQRLLE EEDSKLKGGK APRVATSSKV Sequence homology: Cow: 100%; Dog: 100%; Goat: 92%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rat: 100%; Yeast: 90%.

Required for proper progression of late cytokinetic stages.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCDC124
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Ccdc124
Gene Accession No. Q96CT7
Gene Alias 1810023B24Rik; Ccdc124; coiled-coil domain containing 124; coiled-coil domain-containing protein 124; hypothetical protein LOC510778; RGD1310931
Gene Symbols Ccdc124
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human CCDC124 (aa 46-95).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 115098
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.