Learn More
CCDC124 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A301834AT
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Orangutan
The function of this protein remains unknown.Specifications
| CCDC124 | |
| Polyclonal | |
| Unconjugated | |
| CCDC124 | |
| CCDC124 | |
| Between 100 and 150 | |
| 10 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunoprecipitation | |
| 1 mg/ml | |
| phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
| Q96CT7 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 115098 | |
| 4° C | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.